5-Chloro-1-(4-chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%
Supplier: Apollo Scientific
OR33215-1G
OR33215-5G
APOSOR33215-1GEA
269
GBP
APOSOR33215-1G
APOSOR33215-5G
5-Chloro-1-(4-chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%
5-Chloro-1-(4-chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Formula:
C₁₃H₉Cl₂NO₃ MW: 298,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00140291 CAS Number: 339024-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...