2-Methylisophthalonitrile 95%

Supplier: Apollo Scientific

Synonyms: 2-Methyl-1,3-dicyanobenzene, 2-Methyl-1,3-benzenedicarbonitrile, 2-Methylbenzene-1,3-dicarbonitrile, 2,6-Dicyanotoluene

OR33213-1G OR33213-5G
APOSOR33213-1GEA 159 GBP
APOSOR33213-1G APOSOR33213-5G
2-Methylisophthalonitrile 95%
2-Methylisophthalonitrile
Formula: C₉H₆N₂
MW: 142,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012285
CAS Number: 2317-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR