3-(4-Bromophenyl)-2-propen-1-ol 95%
Supplier: Apollo Scientific
OR33197-1G
OR33197-5G
APOSOR33197-1GEA
211
GBP
APOSOR33197-1G
APOSOR33197-5G
3-(4-Bromophenyl)-2-propen-1-ol 95%
3-(4-Bromophenyl)-2-propen-1-ol
Formula:
C₉H₉BrO MW: 213,08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08056664 CAS Number: 105515-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...