1-(3,5-Dimethyl-1H-pyrazol-1-yl)acetone 95%

Supplier: Apollo Scientific

Synonyms: 1-(3,5-Dimethyl-1H-pyrazol-1-yl)propan-2-one

OR33161-1G OR33161-5G
APOSOR33161-1GEA 139 GBP
APOSOR33161-1G APOSOR33161-5G
1-(3,5-Dimethyl-1H-pyrazol-1-yl)acetone 95%
1-(3,5-Dimethyl-1H-pyrazol-1-yl)acetone
Formula: C₈H₁₂N₂O
MW: 152,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03305776
CAS Number: 361343-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR