N-Acetoacetylmorpholine 95%

Supplier: Apollo Scientific

Synonyms: 1-Morpholinobutane-1,3-dione, 1-(Morpholin-4-yl)butane-1,3-dione

OR32969-1G OR32969-5G
APOSOR32969-1GEA 296 GBP
APOSOR32969-1G APOSOR32969-5G
N-Acetoacetylmorpholine 95%
N-Acetoacetylmorpholine
Formula: C₈H₁₃NO₃
MW: 171,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059774
CAS Number: 16695-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR