Acetophenone oxime 95%
Supplier: Apollo Scientific
Synonyms:
Methyl phenyl ketoxime, 1-Phenyl-1-ethanone oxime
OR32960-25G
OR32960-5G
APOSOR32960-25GEA
820
GBP
APOSOR32960-25G
APOSOR32960-5G
Acetophenone oxime 95%
Acetophenone oxime
Formula:
C₈H₉NO MW: 135,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013931 CAS Number: 613-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...