5-(Chloromethyl)-3-phenylisoxazole 95%

Supplier: Apollo Scientific

OR32869-1G OR32869-5G
APOSOR32869-1GEA 400 GBP
APOSOR32869-1G APOSOR32869-5G
5-(Chloromethyl)-3-phenylisoxazole 95%
5-(Chloromethyl)-3-phenylisoxazole
Formula: C₁₀H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD01935949
CAS Number: 1011-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR