1-(3-Chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%

Supplier: Apollo Scientific

Synonyms: 1-[(3-Chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid

OR32831-1G OR32831-5G OR32831-25G
APOSOR32831-1GEA 159 GBP
APOSOR32831-1G APOSOR32831-5G APOSOR32831-25G
1-(3-Chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%
1-(3-Chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Formula: C₁₃H₁₀ClNO₃
MW: 263,68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00139984
CAS Number: 338754-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR