2-Methyl-2-morpholinopropanal 95%
Supplier: Apollo Scientific
OR32800-1G
OR32800-5G
APOSOR32800-1GEA
400
GBP
APOSOR32800-1G
APOSOR32800-5G
2-Methyl-2-morpholinopropanal 95%
2-Methyl-2-morpholinopropanal
Formula:
C₈H₁₅NO₂ MW: 157,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05663791 CAS Number: 16042-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...