3-Bromo-4-oxo-4-phenylbutanoic acid 95%

Supplier: Apollo Scientific

OR32751-1G OR32751-5G
APOSOR32751-1GEA 151 GBP
APOSOR32751-1G APOSOR32751-5G
3-Bromo-4-oxo-4-phenylbutanoic acid 95%
3-Bromo-4-oxo-4-phenylbutanoic acid
Formula: C₁₀H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD00086369
CAS Number: 53515-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR