1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol 95%
Supplier: Apollo Scientific
OR32716-1G
OR32716-5G
APOSOR32716-1GEA
75.9
GBP
APOSOR32716-1G
APOSOR32716-5G
1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol 95%
1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol
Formula:
C₉H₈ClN₃O MW: 209.633 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00140928 CAS Number: 133902-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...