6-(4-Bromophenyl)imidazo[2,1-b]thiazole 95%
Supplier: Apollo Scientific
OR32674-1G
OR32674-5G
APOSOR32674-1GEA
126
GBP
APOSOR32674-1G
APOSOR32674-5G
6-(4-Bromophenyl)imidazo[2,1-b]thiazole 95%
6-(4-Bromophenyl)imidazo[2,1-b]thiazole
Formula:
C₁₁H₇BrN₂S Storage Temperature: Ambient |
MDL Number:
MFCD00269210 CAS Number: 7120-13-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...