2-(1H-Pyrrol-1-yl)-1-ethanamine, Technical Grade

Supplier: Apollo Scientific

Synonyms: 2-(1H-Pyrrol-1-yl)ethanamine

OR32663-1G OR32663-5G OR32663-25G
APOSOR32663-1GEA 159 GBP
APOSOR32663-1G APOSOR32663-5G APOSOR32663-25G
2-(1H-Pyrrol-1-yl)-1-ethanamine, Technical Grade
2-(1H-Pyrrol-1-yl)-1-ethanamine
Formula: C₆H₁₀N₂
MW: 110,16 g/mol
Density: 1,013 g/cm³ (25 °C)
Flash Pt: 83,5 °C
Storage Temperature: Ambient
MDL Number: MFCD00958089
CAS Number: 29709-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR