1-Phenyl-1H-imidazole-2-thiol 95%
Supplier: Apollo Scientific
OR32630-1G
OR32630-25G
OR32630-5G
APOSOR32630-1GEA
112
GBP
APOSOR32630-1G
APOSOR32630-25G
APOSOR32630-5G
1-Phenyl-1H-imidazole-2-thiol 95%
1-Phenyl-1H-imidazole-2-thiol
Formula:
C₉H₈N₂S Storage Temperature: Ambient |
MDL Number:
MFCD00127929 CAS Number: 17452-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...