7-Chloro-2H-[1,4]benzothiazin-3(4H)-one 95%

Supplier: Apollo Scientific

OR32602-1G OR32602-5G
APOSOR32602-1GEA 145 GBP
APOSOR32602-1G APOSOR32602-5G
7-Chloro-2H-[1,4]benzothiazin-3(4H)-one 95%
7-Chloro-2H-[1,4]benzothiazin-3(4H)-one
Formula: C₈H₆ClNOS
MW: 199,67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00127344
CAS Number: 5333-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR