2-Phenylthiazole-5-carbaldehyde 95%
Supplier: Apollo Scientific
OR32591-25G
OR32591-5G
APOSOR32591-25GEA
709
GBP
APOSOR32591-25G
APOSOR32591-5G
2-Phenylthiazole-5-carbaldehyde 95%
2-Phenylthiazole-5-carbaldehyde
Formula:
C₁₀H₇NOS MW: 189,2300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04973719 CAS Number: 1011-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...