3-Methyl-3,4-dihydro-2(1H)-quinoxalinone 95%

Supplier: Apollo Scientific

OR32541-1G OR32541-5G OR32541-25G
APOSOR32541-1GEA 159 GBP
APOSOR32541-1G APOSOR32541-5G APOSOR32541-25G
3-Methyl-3,4-dihydro-2(1H)-quinoxalinone 95%
3-Methyl-3,4-dihydro-2(1H)-quinoxalinone
Formula: C₉H₁₀N₂O
MW: 162,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00231317
CAS Number: 34070-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR