2-(4-Benzylpiperazino)-2-methylpropanal 95%

Supplier: Apollo Scientific

OR32501-1G OR32501-5G
APOSOR32501-1GEA 400 GBP
APOSOR32501-1G APOSOR32501-5G
2-(4-Benzylpiperazino)-2-methylpropanal 95%
2-(4-Benzylpiperazino)-2-methylpropanal
Formula: C₁₅H₂₂N₂O
MW: 246,36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663785
CAS Number: 886360-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR