4-Chloro-2-oxo-2H-chromene-3-carbaldehyde 95%
Supplier: Apollo Scientific
OR32488-1G
OR32488-25G
OR32488-5G
APOSOR32488-1GEA
145
GBP
APOSOR32488-1G
APOSOR32488-25G
APOSOR32488-5G
4-Chloro-2-oxo-2H-chromene-3-carbaldehyde 95%
4-Chloro-2-oxo-2H-chromene-3-carbaldehyde
Formula:
C₁₀H₅ClO₃ MW: 208,6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00179896 CAS Number: 50329-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...