2-Piperidinecarboxamide 95%

Supplier: Apollo Scientific

Synonyms: Piperidine-2-carboxamide

OR32442-1G OR32442-5G
APOSOR32442-1GEA 57.7 GBP
APOSOR32442-1G APOSOR32442-5G
2-Piperidinecarboxamide 95%
2-Piperidinecarboxamide
Formula: C₆H₁₂N₂O
MW: 128,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663784
CAS Number: 19889-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR