3-(4-Methylpiperazin-1-yl)-3-oxopropanenitrile 95%

Supplier: Apollo Scientific

OR32335-1G OR32335-5G
APOSOR32335-1GEA 139 GBP
APOSOR32335-1G APOSOR32335-5G
3-(4-Methylpiperazin-1-yl)-3-oxopropanenitrile 95%
3-(4-Methylpiperazin-1-yl)-3-oxopropanenitrile
Formula: C₈H₁₃N₃O
Storage Temperature: Ambient
MDL Number: MFCD02186990
CAS Number: 15029-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR