(2-Bromopropyl)benzene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Bromo-1-phenylpropane, (2-Bromopropyl)benzene
OR323250-25G
OR323250-5G
APOSOR323250-25GEA
748
GBP
APOSOR323250-25G
APOSOR323250-5G
(2-Bromopropyl)benzene
(2-Bromopropyl)benzene
Formula:
C₆H₅CH₂CHBrCH₃ MW: 199,09 g/mol Boiling Pt: 107…109 °C Density: 1,29 g/cm³ (25 °C) Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000149 CAS Number: 2114-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...