2-Aminothiazole-5-carbonitrile 97%

Supplier: Apollo Scientific
Warning

OR3230-1G OR3230-250MG
APOSOR3230-1GEA 84 GBP
APOSOR3230-1G APOSOR3230-250MG
2-Aminothiazole-5-carbonitrile 97%
2-Aminothiazole-5-carbonitrile
Formula: C₄H₃N₃S
MW: 125,20 g/mol
Melting Pt: 204…208 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08275724
CAS Number: 51640-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR