N-(tert-Butoxycarbonyl)-L-serinemethylester

Supplier: Apollo Scientific

Synonyms: Boc-Ser-OMe, N-Boc-L-serine Methyl Ester, BOC-L-Serine methyl ester

OR322570-100G OR322570-25G OR322570-500G
APOSOR322570-100GEA 115 GBP
APOSOR322570-100G APOSOR322570-25G APOSOR322570-500G
N-(tert-Butoxycarbonyl)-L-serinemethylester
N-(tert-Butoxycarbonyl)-L-serine methyl ester
Formula: C₉H₁₇NO₅
MW: 219,24 g/mol
Density: 1,082 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00191869
CAS Number: 2766-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR