N-Ethylglycine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(Ethylamino)acetic acid, N-Et-Gly-OH
OR322207-1G
OR322207-5G
APOSOR322207-1GEA
61.6
GBP
APOSOR322207-1G
APOSOR322207-5G
N-Ethylglycine
N-Ethylglycine
Formula:
C₄H₉NO₂ Melting Pt: 190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037794 CAS Number: 627-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...