tert-Butyl carbazate

Supplier: Apollo Scientific
Warning

Synonyms: tert-Butyl hydrazinecarboxylate

OR322134-25G OR322134-100G
APOSOR322134-25GEA 13.2 GBP
APOSOR322134-25G APOSOR322134-100G
tert-Butyl carbazate
tert-Butyl carbazate
Formula: C₅H₁₂N₂O₂
MW: 132,16 g/mol
Boiling Pt: 63…65 °C
Melting Pt: 39…42 °C
Flash Pt: 92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007593
CAS Number: 870-46-2
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR