D-(-)-Quinic acid

Supplier: Apollo Scientific
Warning

Synonyms: (1α,3R,4α,5R)-1,3,4,5-Tetrahydroxycyclohexanecarboxylic acid

OR322105-100G OR322105-250G OR322105-50G
APOSOR322105-100GEA 216 GBP
APOSOR322105-100G APOSOR322105-250G APOSOR322105-50G
D-(-)-Quinic acid
D(-)-Quinic acid
Formula: C₇H₁₂O₆
MW: 192,17 g/mol
Melting Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00003864
CAS Number: 77-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR