5-Iodosalicylic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Hydroxy-5-iodobenzoic acid
OR322048-500G
OR322048-100G
OR322048-25G
APOSOR322048-500GEA
0
GBP
APOSOR322048-500G
APOSOR322048-100G
APOSOR322048-25G
5-Iodosalicylic acid
5-Iodosalicylic acid
Formula:
C₇H₅IO₃ MW: 264,02 g/mol Melting Pt: 194…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002458 CAS Number: 119-30-2 EINECS: 204-313-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...