2-Isopropylphenol
Supplier: Apollo Scientific
|
Danger
|
OR322047-50G
OR322047-250G
OR322047-1KG
APOSOR322047-50GEA
95.7
GBP
APOSOR322047-50G
APOSOR322047-250G
APOSOR322047-1KG
2-Isopropylphenol
2-Isopropylphenol
Formula:
C₉H₁₂O MW: 136,19 g/mol Boiling Pt: 212…213 °C Melting Pt: 42705 °C Density: 1,012 g/cm³ (25 °C) Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002224 CAS Number: 88-69-7 UN: 3145 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...