Methyl-4-chlorocinnamate 98%

Supplier: Apollo Scientific

Synonyms: Methyl 3-(4-chlorophenyl)acrylate, 4-Chlorocinnamic Acid Methyl Ester

OR322013-100G OR322013-25G OR322013-5G
APOSOR322013-100GEA 270 GBP
APOSOR322013-100G APOSOR322013-25G APOSOR322013-5G
Methyl-4-chlorocinnamate 98%
Methyl-4-chlorocinnamate
Formula: C₁₀H₉ClO₂
MW: 196,63 g/mol
Melting Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00075790
CAS Number: 7560-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR