[3-(4-Chlorophenyl)-5-isoxazolyl]methanol 95%
Supplier: Apollo Scientific
Synonyms:
(3-(4-Chlorophenyl)isoxazol-5-yl)methanol
OR32075-1G
OR32075-5G
APOSOR32075-1GEA
269
GBP
APOSOR32075-1G
APOSOR32075-5G
[3-(4-Chlorophenyl)-5-isoxazolyl]methanol 95%
[3-(4-Chlorophenyl)-5-isoxazolyl]methanol
Formula:
C₁₀H₈ClNO₂ MW: 209,6290 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01935957 CAS Number: 206055-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...