7-Bromo-5-nitroindoline 95+%

Supplier: Apollo Scientific

OR318110-1G OR318110-5G
APOSOR318110-1GEA 34.1 GBP
APOSOR318110-1G APOSOR318110-5G
7-Bromo-5-nitroindoline 95+%
7-Bromo-5-nitroindoline
Formula: C₈H₇BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09258800
CAS Number: 87240-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR