2,3,4,5-Tetrahydro-1H-3-benzazepine 90+%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2,3,4,5-Tetrahydro-1H-benzo[d]azepine
OR318069-1G
OR318069-5G
OR318069-500MG
APOSOR318069-1GEA
174
GBP
APOSOR318069-1G
APOSOR318069-5G
APOSOR318069-500MG
2,3,4,5-Tetrahydro-1H-3-benzazepine 90+%
2,3,4,5-Tetrahydro-1H-3-benzazepine
Formula:
C₁₀H₁₃N MW: 147,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04012620 CAS Number: 4424-20-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...