Ethyl-2-cyclohexyl-2-oxoacetate 97%
Supplier: Apollo Scientific
|
Warning
|
OR318068-1G
OR318068-5G
APOSOR318068-1GEA
30.2
GBP
APOSOR318068-1G
APOSOR318068-5G
Ethyl-2-cyclohexyl-2-oxoacetate 97%
Ethyl-2-cyclohexyl-2-oxoacetate
Formula:
C₁₀H₁₆O₃ Storage Temperature: Ambient |
MDL Number:
MFCD09055348 CAS Number: 13275-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...