4-(Methoxycarbonyl)nicotinic acid 90%
Supplier: Apollo Scientific
|
Warning
|
OR318041-1G
OR318041-5G
OR318041-10G
APOSOR318041-1GEA
233
GBP
APOSOR318041-1G
APOSOR318041-5G
APOSOR318041-10G
4-(Methoxycarbonyl)nicotinic acid 90%
4-(Methoxycarbonyl)nicotinic acid
Formula:
C₈H₇NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00111513 CAS Number: 24202-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...