3-(Benzyloxy)cyclobutanol 98%

Supplier: Apollo Scientific
Warning

OR317118-1G OR317118-250MG
APOSOR317118-1GEA 283 GBP
APOSOR317118-1G APOSOR317118-250MG
3-(Benzyloxy)cyclobutanol 98%
3-(Benzyloxy)cyclobutanol
Formula: C₁₁H₁₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD21756137
CAS Number: 100058-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR