4-Bromoisophthalic acid 97%
Supplier: Apollo Scientific
|
Warning
|
OR3158-25G
OR3158-5G
APOSOR3158-25GEA
148
GBP
APOSOR3158-25G
APOSOR3158-5G
4-Bromoisophthalic acid 97%
4-Bromoisophthalic acid
Formula:
C₈H₅BrO₄ MW: 245,03 g/mol Melting Pt: 296…302 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002404 CAS Number: 6939-93-1 EINECS: 230-078-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...