Benzene-1,3,5-tricarbonyl trichloride
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1,3,5-Benzenetricarboxylic acid chloride, 1,3,5-Benzenetricarbonyl chloride, Trimesoylchloride, Trimesoyl chloride
OR315772-100G
OR315772-25G
OR315772-500G
APOSOR315772-100GEA
66.9
GBP
APOSOR315772-100G
APOSOR315772-25G
APOSOR315772-500G
Benzene-1,3,5-tricarbonyl trichloride
Benzene-1,3,5-tricarbonyl trichloride
Formula:
C₉H₃Cl₃O₃ MW: 265,48 g/mol Boiling Pt: 180 °C Melting Pt: 34…38 °C Density: 1,487 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000679 CAS Number: 4422-95-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...