2-(3-Methoxyphenethyl)phenol 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-[2-(3-Methoxyphenyl)ethyl]phenol
OR315766-1G
OR315766-5G
APOSOR315766-1GEA
72.2
GBP
APOSOR315766-1G
APOSOR315766-5G
2-(3-Methoxyphenethyl)phenol 98%
2-(3-Methoxyphenethyl)phenol
Formula:
C₁₅H₁₆O₂ MW: 228,29 g/mol Melting Pt: 47 °C Storage Temperature: Ambient |
MDL Number:
MFCD09800498 CAS Number: 167145-13-3 EINECS: 605-468-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...