4-Phenylbenzyl bromide
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
4-(Bromomethyl)biphenyl, 4-(Bromomethyl)-1,1'-biphenyl, p-(Bromomethyl)biphenyl
OR315762-10G
OR315762-25G
OR315762-5G
APOSOR315762-10GEA
74.8
GBP
APOSOR315762-10G
APOSOR315762-25G
APOSOR315762-5G
4-Phenylbenzyl bromide
4-Phenylbenzyl bromide
Formula:
C₁₃H₁₁Br MW: 247,13 g/mol Boiling Pt: 140 °C/10mmHg Melting Pt: 83…86 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017869 CAS Number: 2567-29-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...