(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone 97%

Supplier: Apollo Scientific

Synonyms: (4R,5S)-4-Methyl-5-phenyl-2-oxazolidone

OR315293-1G OR315293-5G
APOSOR315293-1GEA 55.1 GBP
APOSOR315293-1G APOSOR315293-5G
(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone 97%
(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone
Formula: C₁₀H₁₁NO₂
MW: 177,2000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010845
CAS Number: 77943-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR