Caronic anhydride

Supplier: Apollo Scientific

Synonyms: 3,3-Dimethyl-1,2-cyclopropanedicarboxylic Anhydride, 6,6-Dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione

OR315033-100G OR315033-25G OR315033-5G
APOSOR315033-100GEA 90.5 GBP
APOSOR315033-100G APOSOR315033-25G APOSOR315033-5G
Caronic anhydride
Caronic anhydride
Formula: C₇H₈O₃
MW: 140,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09751198
CAS Number: 67911-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR