4-(Benzyloxy)-4-oxobutanoic acid

Supplier: Apollo Scientific

Synonyms: Succinic acid monobenzyl ester

OR313009-1G OR313009-5G
APOSOR313009-1GEA 46 GBP
APOSOR313009-1G APOSOR313009-5G
4-(Benzyloxy)-4-oxobutanoic acid
4-(Benzyloxy)-4-oxobutanoic acid
Formula: C₁₁H₁₂O₄
MW: 208,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00447717
CAS Number: 103-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR