(S)-(+)-Phenylsuccinic acid ≥99%, ee 99%

Supplier: Apollo Scientific

Synonyms: (S)-2-Phenylsuccinic acid, (S)-(+)-Phenyl-1,4-butanedioic acid

OR310969-1G OR310969-500MG
APOSOR310969-1GEA 145 GBP
APOSOR310969-1G APOSOR310969-500MG
(S)-(+)-Phenylsuccinic acid ≥99%, ee 99%
(S)-(+)-Phenylsuccinic acid
Formula: C₁₀H₁₀O₄
MW: 194,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065929
CAS Number: 4036-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR