Phenylboronic acid neopentyl glycol ester

Supplier: Apollo Scientific

OR310877-25G OR310877-5G
APOSOR310877-25GEA 0 GBP
APOSOR310877-25G APOSOR310877-5G
Phenylboronic acid neopentyl glycol ester
Phenylboronic acid neopentyl glycol ester
Formula: C₁₁H₁₅BO₂
Storage Temperature: Ambient
MDL Number: MFCD05663851
CAS Number: 5123-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR