Methyl cyclopentanecarboxylate 97%

Supplier: Apollo Scientific

OR310647-5G OR310647-25G
APOSOR310647-5GEA 0 GBP
APOSOR310647-5G APOSOR310647-25G
Methyl cyclopentanecarboxylate 97%
Methyl cyclopentanecarboxylate
Formula: C₇H₁₂O₂
Boiling Pt: 158 °C
Density: 0,98 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00019290
CAS Number: 4630-80-2
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR