1,4-Diazabicyclo[2.2.2]octanebis(sulphur dioxide)adduct 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
DABSO, 1,4-Diazabicyclo[2.2.2]octane-1,4-diium-1,4-disulphinate, DABCO-Bis(sulphur dioxide), Bis(sulphur dioxide)-1,4-diazabicyclo[2.2.2]octane
OR310371-1G
OR310371-5G
APOSOR310371-1GEA
13.2
GBP
APOSOR310371-1G
APOSOR310371-5G
1,4-Diazabicyclo[2.2.2]octanebis(sulphur dioxide)adduct 97%
1,4-Diazabicyclo[2.2.2]octanebis(sulphur dioxide)adduct
Formula:
C₆H₁₂N₂O₄S₂ MW: 240,29 g/mol Melting Pt: 168…172 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD22377802 CAS Number: 119752-83-9 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...