tert-Butyl (5-bromothiazol-2-yl)carbamate

Supplier: Apollo Scientific
Warning

OR310323-1G OR310323-5G
APOSOR310323-1GEA 51.2 GBP
APOSOR310323-1G APOSOR310323-5G
tert-Butyl (5-bromothiazol-2-yl)carbamate
tert-Butyl (5-bromothiazol-2-yl)carbamate
Formula: C₈H₁₁BrN₂O₂S
MW: 279,15 g/mol
Melting Pt: 149 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368614
CAS Number: 405939-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR