1-(4-Bromophenyl)pyrrolidine

Supplier: Apollo Scientific
Warning

Synonyms: N-(4-Bromophenyl)pyrrolidine

OR31021-1G OR31021-5G OR31021-10G
APOSOR31021-1GEA 27.6 GBP
APOSOR31021-1G APOSOR31021-5G APOSOR31021-10G
1-(4-Bromophenyl)pyrrolidine
1-(4-Bromophenyl)pyrrolidine
Formula: C₁₀H₁₂BrN
MW: 226,1130 g/mol
Melting Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD04112480
CAS Number: 22090-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR