2-Chloro-3,5-diiodopyridine
Supplier: Apollo Scientific
OR310200-1G
OR310200-500MG
APOSOR310200-1GEA
551
GBP
APOSOR310200-1G
APOSOR310200-500MG
2-Chloro-3,5-diiodopyridine
2-Chloro-3,5-diiodopyridine
Formula:
C₅H₂ClI₂N MW: 365,34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04966987 CAS Number: 856169-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...